Tisk…

(S)-tert-Butyl 2-amino-3-methylbutylcarbamate 97%

Dodavatel: Apollo Scientific
Varování

OR452193-1G OR452193-250MG OR452193-500MG
APOSOR452193-1GEA 29760 CZK
APOSOR452193-1G APOSOR452193-250MG APOSOR452193-500MG
(S)-tert-Butyl 2-amino-3-methylbutylcarbamate 97%
(S)-tert-Butyl 2-amino-3-methylbutylcarbamate
Vzorec: C₁₀H₂₂N₂O₂
MW: 202,294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18694786
CAS číslo: 400652-55-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR