5-Acetyl-2-bromo-3-(cyclopropyl)thiophene 95%
Dodavatel: Apollo Scientific
OR45219-1G
OR45219-250MG
APOSOR45219-1GEA
16340
CZK
APOSOR45219-1G
APOSOR45219-250MG
5-Acetyl-2-bromo-3-(cyclopropyl)thiophene 95%
5-Acetyl-2-bromo-3-(cyclopropyl)thiophene
Vzorec:
C₉H₉BrOS MW: 245,136 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32219472 CAS číslo: 2302672-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...