Tisk…
(2S)-N1,N1-Dimethyl-1,2-propanediamine dihydrochloride 97%
Dodavatel: Apollo Scientific
OR452188-1G OR452188-250MG OR452188-500MG
APOSOR452188-1GEA 25660 CZK
APOSOR452188-1G APOSOR452188-250MG APOSOR452188-500MG
(2S)-N1,N1-Dimethyl-1,2-propanediamine dihydrochloride 97%
(2S)-N1,N1-Dimethyl-1,2-propanediamine dihydrochloride
Vzorec: C₅H₁₄N₂·2ClH
MW: 175,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619103
CAS číslo: 82105-48-4