a-(1-Aminoethyl)-4-chloro-benzenemethanol hydrochloride 97%
Dodavatel: Apollo Scientific
OR452184-1G
OR452184-250MG
OR452184-500MG
APOSOR452184-1GEA
22320
CZK
APOSOR452184-1G
APOSOR452184-250MG
APOSOR452184-500MG
a-(1-Aminoethyl)-4-chloro-benzenemethanol hydrochloride 97%
a-(1-Aminoethyl)-4-chloro-benzenemethanol hydrochloride
Vzorec:
C₉H₁₂ClNO·ClH MW: 222,112 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09028116 CAS číslo: 23933-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...