b-Amino-2-bromobenzenepropanol 97%
Dodavatel: Apollo Scientific
OR452183-1G
OR452183-250MG
OR452183-500MG
APOSOR452183-1GEA
22320
CZK
APOSOR452183-1G
APOSOR452183-250MG
APOSOR452183-500MG
b-Amino-2-bromobenzenepropanol 97%
b-Amino-2-bromobenzenepropanol
Vzorec:
C₉H₁₂BrNO Storage Temperature: Ambient |
MDL Number:
MFCD11040142 CAS číslo: 1260786-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...