Tisk…

N-(3-Aminopropyl)-N-methyl-acetamide hydrochloride 97%

Dodavatel: Apollo Scientific

OR452181-1G OR452181-250MG OR452181-500MG
APOSOR452181-1GEA 22320 CZK
APOSOR452181-1G APOSOR452181-250MG APOSOR452181-500MG
N-(3-Aminopropyl)-N-methyl-acetamide hydrochloride 97%
N-(3-Aminopropyl)-N-methyl-acetamide hydrochloride
Vzorec: C₆H₁₄N₂O·ClH
MW: 166,649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29991785
CAS číslo: 84598-51-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR