Tisk…

(S)-b-Amino-3-methoxybenzenepropanol 97%

Dodavatel: Apollo Scientific

OR452179-1G OR452179-250MG OR452179-500MG
APOSOR452179-1GEA 24800 CZK
APOSOR452179-1G APOSOR452179-250MG APOSOR452179-500MG
(S)-b-Amino-3-methoxybenzenepropanol 97%
(S)-b-Amino-3-methoxybenzenepropanol
Vzorec: C₁₀H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD17214662
CAS číslo: 938462-27-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR