(S)-b-Amino-3-nitrobenzenepropanol 97%
Dodavatel: Apollo Scientific
OR452178-1G
OR452178-250MG
OR452178-500MG
APOSOR452178-1GEA
24800
CZK
APOSOR452178-1G
APOSOR452178-250MG
APOSOR452178-500MG
(S)-b-Amino-3-nitrobenzenepropanol 97%
(S)-b-Amino-3-nitrobenzenepropanol
Vzorec:
C₉H₁₂N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD17214654 CAS číslo: 792179-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...