Tisk…

b-Amino-3-methoxybenzeneethanol 97%

Dodavatel: Apollo Scientific

OR452172-1G OR452172-250MG OR452172-500MG
APOSOR452172-1GEA 22320 CZK
APOSOR452172-1G APOSOR452172-250MG APOSOR452172-500MG
b-Amino-3-methoxybenzeneethanol 97%
b-Amino-3-methoxybenzeneethanol
Vzorec: C₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD11213674
CAS číslo: 325153-00-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR