b-Amino-3-bromobenzeneethanol 97%
Dodavatel: Apollo Scientific
Synonyms:
2-Amino-2-(3-bromophenyl)ethanol
OR452171-1G
APOSOR452171-1GEA
2070
CZK
APOSOR452171-1G
b-Amino-3-bromobenzeneethanol 97%
b-Amino-3-bromobenzeneethanol
Vzorec:
C₈H₁₀BrNO MW: 216,075 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12827191 CAS číslo: 188586-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...