2-Amino-1-(4-methylphenyl)ethanol hydrochloride 97%
Dodavatel: Apollo Scientific
OR452164-1G
OR452164-500MG
APOSOR452164-1GEA
18020
CZK
APOSOR452164-1G
APOSOR452164-500MG
2-Amino-1-(4-methylphenyl)ethanol hydrochloride 97%
2-Amino-1-(4-methylphenyl)ethanol hydrochloride
Vzorec:
C₉H₁₃NO·ClH MW: 187,667 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09028159 CAS číslo: 53360-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...