Tisk…
2-Amino-1-(3-bromophenyl)ethanol hydrochloride 97%
Dodavatel: Apollo Scientific
OR452163-1G OR452163-500MG
APOSOR452163-1GEA 18620 CZK
APOSOR452163-1G APOSOR452163-500MG
2-Amino-1-(3-bromophenyl)ethanol hydrochloride 97%
2-Amino-1-(3-bromophenyl)ethanol hydrochloride
Vzorec: C₈H₁₀BrNO·ClH
MW: 252,536 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05861636
CAS číslo: 41147-81-3