Tisk…

N-Boc-2-(4-bromophenyl)-2-hydroxyethanamine 97%

Dodavatel: Apollo Scientific

OR452161-1G OR452161-500MG
APOSOR452161-1GEA 19910 CZK
APOSOR452161-1G APOSOR452161-500MG
N-Boc-2-(4-bromophenyl)-2-hydroxyethanamine 97%
N-Boc-2-(4-bromophenyl)-2-hydroxyethanamine
Vzorec: C₁₃H₁₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD08544096
CAS číslo: 913181-90-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR