Tisk…

4-(Cyclopropyl)thiophene-2-carboxylic acid

Dodavatel: Apollo Scientific

OR45216-1G OR45216-250MG OR45216-5G
APOSOR45216-1GEA 12200 CZK
APOSOR45216-1G APOSOR45216-250MG APOSOR45216-5G
4-(Cyclopropyl)thiophene-2-carboxylic acid
4-(Cyclopropyl)thiophene-2-carboxylic acid
Vzorec: C₈H₈O₂S
MW: 168,213 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22067668
CAS číslo: 1252903-38-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR