N-Cbz-2-amino-1-butanol 97%
Dodavatel: Apollo Scientific
OR452152-1G
OR452152-25G
OR452152-5G
APOSOR452152-1GEA
3940
CZK
APOSOR452152-1G
APOSOR452152-25G
APOSOR452152-5G
N-Cbz-2-amino-1-butanol 97%
N-Cbz-2-amino-1-butanol
Vzorec:
C₁₂H₁₇NO₃ MW: 223,268 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12180538 CAS číslo: 137160-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...