Tisk…

N-Cbz-(S)-2-amino-1-butanol 97%

Dodavatel: Apollo Scientific

OR452151-1G OR452151-25G OR452151-5G
APOSOR452151-1GEA 3940 CZK
APOSOR452151-1G APOSOR452151-25G APOSOR452151-5G
N-Cbz-(S)-2-amino-1-butanol 97%
N-Cbz-(S)-2-amino-1-butanol
Vzorec: C₁₂H₁₇NO₃
MW: 223,268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29050124
CAS číslo: 22373-14-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR