Tisk…

N-Boc-(S)-2-amino-1-butanol 97%

Dodavatel: Apollo Scientific

Synonyms: (S)-tert-Butyl (1-hydroxybutan-2-yl)carbamate

OR452149-25G
APOSOR452149-25GEA 3740 CZK
APOSOR452149-25G
N-Boc-(S)-2-amino-1-butanol 97%
N-Boc-(S)-2-amino-1-butanol
Vzorec: C₉H₁₉NO₃
MW: 189,252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974275
CAS číslo: 150736-72-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR