(S)-b-Amino-4-methoxy-benzeneethanol 97%
Dodavatel: Apollo Scientific
Synonyms:
(S)-2-Amino-2-(4-methoxyphenyl)ethanol
OR452142-1G
OR452142-250MG
APOSOR452142-1GEA
25560
CZK
APOSOR452142-1G
APOSOR452142-250MG
(S)-b-Amino-4-methoxy-benzeneethanol 97%
(S)-b-Amino-4-methoxy-benzeneethanol
Vzorec:
C₉H₁₃NO₂ MW: 167.205 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09253728 CAS číslo: 191109-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...