Tisk…

2-Amino-2-(3-methylphenyl)ethan-1-ol 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-2-(m-tolyl)ethanol

OR452140-1G OR452140-250MG
APOSOR452140-1GEA 28730 CZK
APOSOR452140-1G APOSOR452140-250MG
2-Amino-2-(3-methylphenyl)ethan-1-ol 97%
2-Amino-2-(3-methylphenyl)ethan-1-ol
Vzorec: C₉H₁₃NO
MW: 151,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827183
CAS číslo: 1179634-13-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR