Tisk…

Methyl 3-(cyclopropyl)thiophene-2-carboxylate

Dodavatel: Apollo Scientific

OR45214-1G OR45214-250MG
APOSOR45214-1GEA 13460 CZK
APOSOR45214-1G APOSOR45214-250MG
Methyl 3-(cyclopropyl)thiophene-2-carboxylate
Methyl 3-(cyclopropyl)thiophene-2-carboxylate
Vzorec: C₉H₁₀O₂S
MW: 182,24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32219469
CAS číslo: 1871848-03-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR