1-(3-Bromopropyl)-4-methoxybenzene 97%
Dodavatel: Apollo Scientific
OR452133-1G
OR452133-5G
APOSOR452133-1GEA
2200
CZK
APOSOR452133-1G
APOSOR452133-5G
1-(3-Bromopropyl)-4-methoxybenzene 97%
1-(3-Bromopropyl)-4-methoxybenzene
Vzorec:
C₁₀H₁₃BrO Storage Temperature: Ambient |
MDL Number:
MFCD00155066 CAS číslo: 57293-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...