1-Boc-4-(4-chlorobenzyl)-4-carboxypiperidine 97%
Dodavatel: Apollo Scientific
Synonyms:
1-(tert-Butoxycarbonyl)-4-(4-chlorobenzyl)piperidine-4-carboxylic acid
OR452131-1G
OR452131-250MG
APOSOR452131-1GEA
16340
CZK
APOSOR452131-1G
APOSOR452131-250MG
1-Boc-4-(4-chlorobenzyl)-4-carboxypiperidine 97%
1-Boc-4-(4-chlorobenzyl)-4-carboxypiperidine
Vzorec:
C₁₈H₂₄ClNO₄ MW: 353.84 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08461314 CAS číslo: 170284-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...