Tisk…

1-Boc-4,4-bis(hydroxymethyl)piperidine 97%

Dodavatel: Apollo Scientific

OR452121-250MG OR452121-500MG
APOSOR452121-250MGEA 14880 CZK
APOSOR452121-250MG APOSOR452121-500MG
1-Boc-4,4-bis(hydroxymethyl)piperidine 97%
1-Boc-4,4-bis(hydroxymethyl)piperidine
Vzorec: C₁₂H₂₃NO₄
MW: 245,315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12407070
CAS číslo: 374794-84-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR