Methyl 5-(cyclopropyl)thiophene-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR45212-1G
OR45212-250MG
OR45212-5G
APOSOR45212-1GEA
14880
CZK
APOSOR45212-1G
APOSOR45212-250MG
APOSOR45212-5G
Methyl 5-(cyclopropyl)thiophene-2-carboxylate
Methyl 5-(cyclopropyl)thiophene-2-carboxylate
Vzorec:
C₉H₁₀O₂S MW: 182,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32219468 CAS číslo: 1021432-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...