Tisk…

(R)-3-Phenoxypyrrolidine hydrochloride 97%

Dodavatel: Apollo Scientific

OR452100-1G OR452100-500MG
APOSOR452100-1GEA 14880 CZK
APOSOR452100-1G APOSOR452100-500MG
(R)-3-Phenoxypyrrolidine hydrochloride 97%
(R)-3-Phenoxypyrrolidine hydrochloride
Vzorec: C₁₀H₁₃NO·ClH
Storage Temperature: Ambient
MDL Number: MFCD30491355
CAS číslo: 1366664-52-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR