Tisk…

Boc-N-methyl-DL-Leucine 97%

Dodavatel: Apollo Scientific

OR452096-1G OR452096-5G
APOSOR452096-1GEA 5030 CZK
APOSOR452096-1G APOSOR452096-5G
Boc-N-methyl-DL-Leucine 97%
Boc-N-methyl-DL-Leucine
Vzorec: C₁₂H₂₃NO₄
MW: 245,315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00480937
CAS číslo: 13734-32-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR