Tisk…

N-Fmoc-2-(methylamino)butyric acid 97%

Dodavatel: Apollo Scientific

OR452092-1G OR452092-500MG
APOSOR452092-1GEA 14880 CZK
APOSOR452092-1G APOSOR452092-500MG
N-Fmoc-2-(methylamino)butyric acid 97%
N-Fmoc-2-(methylamino)butyric acid
Vzorec: C₂₀H₂₁NO₄
MW: 339.385 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18328457
CAS číslo: 1341092-00-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR