Tisk…

3-(Cyclohexylmethoxy)-benzoic acid 97%

Dodavatel: Apollo Scientific

OR452065-1G OR452065-250MG
APOSOR452065-1GEA 16170 CZK
APOSOR452065-1G APOSOR452065-250MG
3-(Cyclohexylmethoxy)-benzoic acid 97%
3-(Cyclohexylmethoxy)-benzoic acid
Vzorec: C₁₄H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD11122824
CAS číslo: 158860-95-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR