L-Alanine cyclobutylmethyl ester hydrochloride 97%
Dodavatel: Apollo Scientific
OR452064-1G
OR452064-5G
APOSOR452064-1GEA
9690
CZK
APOSOR452064-1G
APOSOR452064-5G
L-Alanine cyclobutylmethyl ester hydrochloride 97%
L-Alanine cyclobutylmethyl ester hydrochloride
Vzorec:
C₈H₁₅NO₂·ClH MW: 193,671 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27952567 CAS číslo: 261909-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...