1-(3-Bromophenyl)cyclopentanecarboxylic acid 97%
Dodavatel: Apollo Scientific
OR452055-1G
OR452055-5G
APOSOR452055-1GEA
1370
CZK
APOSOR452055-1G
APOSOR452055-5G
1-(3-Bromophenyl)cyclopentanecarboxylic acid 97%
1-(3-Bromophenyl)cyclopentanecarboxylic acid
Vzorec:
C₁₂H₁₃BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00573784 CAS číslo: 143328-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...