4-Bromo-DL-homophenylalanine 97%
Dodavatel: Apollo Scientific
OR452037-1G
OR452037-250MG
OR452037-500MG
APOSOR452037-1GEA
18620
CZK
APOSOR452037-1G
APOSOR452037-250MG
APOSOR452037-500MG
4-Bromo-DL-homophenylalanine 97%
4-Bromo-DL-homophenylalanine
Vzorec:
C₁₀H₁₂BrNO₂ MW: 258,112 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07372197 CAS číslo: 225233-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...