Tisk…

2-Amino-3-biphenyl-3-yl-propionic acid 97%

Dodavatel: Apollo Scientific

OR452032-1G OR452032-250MG OR452032-500MG
APOSOR452032-1GEA 18620 CZK
APOSOR452032-1G APOSOR452032-250MG APOSOR452032-500MG
2-Amino-3-biphenyl-3-yl-propionic acid 97%
2-Amino-3-biphenyl-3-yl-propionic acid
Vzorec: C₁₅H₁₅NO₂
MW: 241,285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07372093
CAS číslo: 887502-03-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR