Tisk…

4-Bromo-DL-tryptophan 97%

Dodavatel: Apollo Scientific

OR452026-1G OR452026-250MG OR452026-500MG
APOSOR452026-1GEA 18620 CZK
APOSOR452026-1G APOSOR452026-250MG APOSOR452026-500MG
4-Bromo-DL-tryptophan 97%
4-Bromo-DL-tryptophan
Vzorec: C₁₁H₁₁BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09915904
CAS číslo: 25796-04-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR