Tisk…

(S)-2-Amino-5-methylhex-4-enoic acid 97%

Dodavatel: Apollo Scientific

OR452020-1G OR452020-500MG
APOSOR452020-1GEA 18620 CZK
APOSOR452020-1G APOSOR452020-500MG
(S)-2-Amino-5-methylhex-4-enoic acid 97%
(S)-2-Amino-5-methylhex-4-enoic acid
Vzorec: C₇H₁₃NO₂
MW: 143,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10565736
CAS číslo: 19914-06-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR