Tisk…

4-(Cyclopropyl)thiophene-2-carbonitrile

Dodavatel: Apollo Scientific

OR45202-1G OR45202-250MG
APOSOR45202-1GEA 19840 CZK
APOSOR45202-1G APOSOR45202-250MG
4-(Cyclopropyl)thiophene-2-carbonitrile
4-(Cyclopropyl)thiophene-2-carbonitrile
Vzorec: C₈H₇NS
MW: 149,213 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28059592
CAS číslo: 1823927-97-4
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR