Tisk…

2,6-Dichloro-DL-phenylalanine 97%

Dodavatel: Apollo Scientific

OR452015-1G OR452015-500MG OR452015-5G
APOSOR452015-1GEA 7440 CZK
APOSOR452015-1G APOSOR452015-500MG APOSOR452015-5G
2,6-Dichloro-DL-phenylalanine 97%
2,6-Dichloro-DL-phenylalanine
Vzorec: C₉H₉Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD05228013
CAS číslo: 110300-03-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR