Tisk…

Fmoc-3-Cyclohexylalanine 97%

Dodavatel: Apollo Scientific

OR452012-1G OR452012-5G
APOSOR452012-1GEA 3940 CZK
APOSOR452012-1G APOSOR452012-5G
Fmoc-3-Cyclohexylalanine 97%
Fmoc-3-Cyclohexylalanine
Vzorec: C₂₄H₂₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00274812
CAS číslo: 188632-07-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR