Tisk…

3-Iodo-DL-phenylalanine 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-3-(3-iodophenyl)propanoic acid, H-DL-Phe(3-I)-OH

OR452004-1G OR452004-5G
APOSOR452004-1GEA 4370 CZK
APOSOR452004-1G APOSOR452004-5G
3-Iodo-DL-phenylalanine 97%
3-Iodo-DL-phenylalanine
Vzorec: C₉H₁₀INO₂
MW: 291,086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05227974
CAS číslo: 20846-38-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR