Tisk…

2-Iodo-DL-phenylalanine 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-3-(2-iodophenyl)propanoic acid, H-DL-Phe(2-I)-OH

OR452003-1G OR452003-500MG OR452003-5G OR452003-250MG
APOSOR452003-1GEA 5720 CZK
APOSOR452003-1G APOSOR452003-500MG APOSOR452003-5G APOSOR452003-250MG
2-Iodo-DL-phenylalanine 97%
2-Iodo-DL-phenylalanine
Vzorec: C₉H₁₀INO₂
MW: 291.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05227973
CAS číslo: 1986-86-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR