Tisk…

2-Cyano-DL-phenylalanine 97%

Dodavatel: Apollo Scientific

OR452002-1G OR452002-5G
APOSOR452002-1GEA 3940 CZK
APOSOR452002-1G APOSOR452002-5G
2-Cyano-DL-phenylalanine 97%
2-Cyano-DL-phenylalanine
Vzorec: C₁₀H₁₀N₂O₂
MW: 190.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264319
CAS číslo: 263396-40-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR