Tisk…

4-(Cyclopropyl)thiophene-2-carboxaldehyde

Dodavatel: Apollo Scientific

OR45199-1G OR45199-250MG
APOSOR45199-1GEA 22520 CZK
APOSOR45199-1G APOSOR45199-250MG
4-(Cyclopropyl)thiophene-2-carboxaldehyde
4-(Cyclopropyl)thiophene-2-carboxaldehyde
Vzorec: C₈H₈OS
MW: 152,214 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06802673
CAS číslo: 893738-62-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR