2-Methylbenzofuran-6-ol
Dodavatel: Apollo Scientific
OR451800-100MG
OR451800-250MG
APOSOR451800-100MGEA
1820
CZK
APOSOR451800-100MG
APOSOR451800-250MG
2-Methylbenzofuran-6-ol
2-Methylbenzofuran-6-ol
Vzorec:
C₉H₈O₂ MW: 148,159 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09908136 CAS číslo: 54584-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...