N-(3-Bromophenyl)cyanamide
Dodavatel: Apollo Scientific
|
Varování
|
OR45179-1G
OR45179-250MG
APOSOR45179-1GEA
7910
CZK
APOSOR45179-1G
APOSOR45179-250MG
N-(3-Bromophenyl)cyanamide
N-(3-Bromophenyl)cyanamide
Vzorec:
C₇H₅BrN₂ MW: 197,032 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16767982 CAS číslo: 70590-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...