Tisk…

2-Cyclopropylbenzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR45172-1G OR45172-250MG OR45172-5G
APOSOR45172-1GEA 3810 CZK
APOSOR45172-1G APOSOR45172-250MG APOSOR45172-5G
2-Cyclopropylbenzenesulphonyl chloride
2-Cyclopropylbenzenesulphonyl chloride
Vzorec: C₉H₉ClO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD31666885
CAS číslo: 1772586-39-6
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR