Tisk…

Ethyl 4-(2-oxoimidazolidin-1-yl)benzoate

Dodavatel: Apollo Scientific
Varování

OR45171-1G OR45171-250MG
APOSOR45171-1GEA 2900 CZK
APOSOR45171-1G APOSOR45171-250MG
Ethyl 4-(2-oxoimidazolidin-1-yl)benzoate
Ethyl 4-(2-oxoimidazolidin-1-yl)benzoate
Vzorec: C₁₂H₁₄N₂O₃
MW: 234,251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066839
CAS číslo: 167626-44-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR