7-Bromobenzofuran-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR45170-1G
OR45170-250MG
APOSOR45170-1GEA
4140
CZK
APOSOR45170-1G
APOSOR45170-250MG
7-Bromobenzofuran-2-carboxylic acid
7-Bromobenzofuran-2-carboxylic acid
Vzorec:
C₉H₅BrO₃ Storage Temperature: Ambient |
MDL Number:
MFCD10699647 CAS číslo: 550998-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...