Ethyl 7-bromo-5-methoxy-1-benzofuran-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR45165-1G
OR45165-250MG
APOSOR45165-1GEA
4140
CZK
APOSOR45165-1G
APOSOR45165-250MG
Ethyl 7-bromo-5-methoxy-1-benzofuran-2-carboxylate
Ethyl 7-bromo-5-methoxy-1-benzofuran-2-carboxylate
Vzorec:
C₁₂H₁₁BrO₄ Storage Temperature: Ambient |
MDL Number:
MFCD31726338 CAS číslo: 91571-13-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...