Ethyl 5-bromo-7-methoxy-1-benzofuran-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR45163-1G
OR45163-250MG
APOSOR45163-1GEA
3640
CZK
APOSOR45163-1G
APOSOR45163-250MG
Ethyl 5-bromo-7-methoxy-1-benzofuran-2-carboxylate
Ethyl 5-bromo-7-methoxy-1-benzofuran-2-carboxylate
Vzorec:
C₁₂H₁₁BrO₄ MW: 299,117 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16089983 CAS číslo: 150612-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...