Tisk…

4-Iodo-1-methyl-1H-pyrazol-3-amine

Dodavatel: Apollo Scientific
Nebezpečí

OR451431-1G OR451431-25G OR451431-5G
APOSOR451431-1GEA 7610 CZK
APOSOR451431-1G APOSOR451431-25G APOSOR451431-5G
4-Iodo-1-methyl-1H-pyrazol-3-amine
4-Iodo-1-methyl-1H-pyrazol-3-amine
Vzorec: C₄H₆IN₃
MW: 223,015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00847646
CAS číslo: 150187-16-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR