Tisk…

4-Methyl-1H-pyrazol-3-amine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR451429-1G OR451429-5G
APOSOR451429-1GEA 10750 CZK
APOSOR451429-1G APOSOR451429-5G
4-Methyl-1H-pyrazol-3-amine hydrochloride
4-Methyl-1H-pyrazol-3-amine hydrochloride
Vzorec: C₄H₇N₃·ClH
MW: 133,579 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25368943
CAS číslo: 1431963-06-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR